Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Recordall™ Chart Paper
Chart Paper for Discontinued Fisher Recordall™ Series 5000 Recorders
Description
- Roll is 11 in. wide (28cm) and 100 ft. long (30m)
- Major Division is numbered along the amplitude axis for all units except Cat. No. 13-939-37
Specifications
Continuous Record Chart Paper | |
For Fisher Recordall™ series 5000 recorders | |
30.48 m |
Major divisions every 1 in.; 5 minor divisions/major | |
100 ft. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction