Learn More
Fisherbrand™ Red-Spirit™ Fluoropolymer-Coated Laboratory Thermometers
Precision grade with white back
Description
- Transparent coating of PTFE resin prevents shattering
- For 76mm immersion only
- Very accurate temperature readings without the environmental hazards of mercury
- Thermometers with low ranges of -200°C to +30°C are filled with pentane; -100°C to +50°C with xylene mixture
- Red liquid thermometers at other ranges are filled with kerosene
Specifications
76 mm | |
0°F | |
0°F to 230°F |
230°F | |
2°F |