Learn More
Fisherbrand™ Red-Spirit™ Fluoropolymer-Coated Laboratory Thermometers
Precision grade with white back
Products 1
Catalog Number Description Price Qty Availability  
Catalog Number Description Price Qty Availability  
14-983-25A
View Documents
N/A Range: 0° to +230°F This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Transparent coating of PTFE resin prevents shattering
  • For 76mm immersion only
  • Very accurate temperature readings without the environmental hazards of mercury
  • Thermometers with low ranges of -200°C to +30°C are filled with pentane; -100°C to +50°C with xylene mixture
  • Red liquid thermometers at other ranges are filled with kerosene