Learn More
Fisherbrand™ Red-Spirit™ General-Purpose Laboratory Thermometers
Very accurate temperature readings without the environmental hazards of mercury
Catalog No. 15-038
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
  • Feature white backing
  • Multiple calibration points are used to compensate for nonlinear expansion of liquid
  • Thermometers with low ranges of -200°C to +30°C are filled with pentane; -100°C to +50°C with xylene mixture
  • Red liquid thermometers at other ranges are filled with kerosene
Liquid in the 15-038 thermometer will begin to freeze at about -193°C, but only after 30 to 60 minutes of exposure. Indicated immersion levels must be observed for accurate readings.
Certifications
NIST traceable, Accuracy to ANSI™/SAMA tolerances
Specifications
Specifications
Total
350 mm
−200°C
1°C
13.77 in.
30°C
-200°C to +30°C