Learn More
Fisherbrand™ Red-Spirit™ Pocket Thermometers
Filled with kerosene and nontoxic red dye
 View more versions of this product
Catalog No. 15-021C
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • With white back and permanently fused markings printed horizontally for easy reading
  • Complete with protective metal case and pocket clip
Store in upright position to avoid column separation.
Specifications
Specifications
-30°F to +120°F
−30°F
Total
152 mm
120°F
2°F
6 in.