Fisherbrand™ Redi-Tip™ Pipet Tips: General Purpose Available on GSA/VA Contract for Federal Government customers only.

Premium virgin polypropylene resin and advanced molding methods produce a superior, high-quality pipet tip

 View more versions of this product

Catalog No. 02-707-502

  null
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Packaging Hinged Rack, 10 x 96
    Sterility Sterile
    Product Type Redi-Tip™ Tip
    Material Polypropylene
    Length (English) 2 in.
    Length (Metric) 5cm
    For Use With (Equipment) Research grade pipetters
    Autoclavable Yes
    Tip Style Hinged Rack
    Certifications/Compliance RNase, DNase, DNA, bioburden, PCR inhibitor, pyrogen-free
    Volume (Metric) 1 to 200μL
    Color Yellow

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit