Fisherbrand™ Redi-Tip™ Pipet Tips: Reference

Premium virgin polypropylene resin and advanced molding methods produce a superior, high-quality pipet tip

 View more versions of this product

Catalog No. 02-707-119

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Product Type Redi-Tip™ Tip
    Material Polypropylene
    Packaging Lift-off Rack, 10 x 96
    Length (English) 2.31 in.
    Length (Metric) 5.87cm
    For Use With (Equipment) Research grade pipetters
    Sterility Sterile
    Autoclavable Yes
    Tip Style Elongated
    Certifications/Compliance RNase, DNase, DNA, bioburden, PCR inhibitor, pyrogen-free
    Volume (Metric) 1 to 300μL
    Color Natural

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit