Learn More
Fisherbrand™ Regenerated Cellulose Dialysis Tubing
Diffuses low molecular weight compounds in aqueous solutions. Fisherbrand™ Regenerated Cellulose Dialysis Tubing retains proteins and bacteria in tubing.
$305.00
Specifications
Material Regenerated Cellulose
Quantity Each
For Use With (Application) Diffuses low molecular weight compounds in aqueous solutions, retains proteins and bacteria in tubing
Kit Contents Two locking nylon membrane clamps, pair of disposable scissors, individual lot data sheets supplied with tubing types T1, T2, and T3
Item Description Dialysis Tubing
View More Specs
Includes: Two locking nylon membrane clamps; pair of disposable scissors; individual lot data sheets supplied with tubing Types T1, T2, and T3
Products 13
Catalog Number Diameter (English) Length (English) MWCO Product Type Price Quantity & Availability  
Catalog Number Diameter (English) Length (English) MWCO Product Type Price Quantity & Availability  
21-152-10
View Documents
0.47 in. 49.2 ft. 3500 d T1 Each for $305.00
Add to cart
21-152-14
View Documents
1.125 in. 98.42 ft. 12000 to 14000 d T4 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
21-152-15
View Documents
0.82 in. 98.42 ft. 12000 to 14000 d T4 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
21-152-4
View Documents
0.8 in. 98.42 ft. 6000 to 8000 d T2 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
21-152-18
View Documents
0.62 in. 49.2 ft. 12000 to 14000 d T3 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
21-152-16
View Documents
0.62 in. 98.42 ft. 12000 to 14000 d T4 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
21-152-3
View Documents
0.57 in. 98.42 ft. 6000 to 8000 d T2 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
21-152-5
View Documents
1 in. 98.42 ft. 6000 to 8000 d T2 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
21-152-6
View Documents
1.25 in. 98.42 ft. 6000 to 8000 d T2 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
21-152-17
View Documents
0.25 in. 98.42 ft. 12000 to 14000 d T4 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
21-152-7
View Documents
0.25 in. 49.2 ft. 12000 to 14000 d T3 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
21-152-8
View Documents
1.125 in. 49.2 ft. 12000 to 14000 d T3 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
21-152-9
View Documents
1.15 in. 49.2 ft. 3500 d T1 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Diffuses low molecular weight compounds in aqueous solutions. Fisherbrand™ Regenerated Cellulose Dialysis Tubing retains proteins and bacteria in tubing.
  • Made from regenerated cellulose membrane
  • MWCOs from 3500 to 14,000 Daltons
  • Individual lot data sheets from outside laboratory supplied with tubing Types T1, T2, and T3; standard grade Type T4 is not tested in outside laboratory
Order Info
Comes on compact polystyrene spool that eliminates adsorption of glycerin from membranes. Dispenser box is packaged in plastic bag with desiccant for humidity control.
Specifications
Specifications
Regenerated Cellulose
Diffuses low molecular weight compounds in aqueous solutions, retains proteins and bacteria in tubing
Dialysis Tubing
Each
Two locking nylon membrane clamps, pair of disposable scissors, individual lot data sheets supplied with tubing types T1, T2, and T3