Learn More
Fisherbrand™ Regenerated Cellulose Dialysis Tubing
Diffuses low molecular weight compounds in aqueous solutions; retains proteins and bacteria in tubing
 View more versions of this product
Catalog No. 21-152-16
  Check Availability
Add to cart
Specifications
Description
  • Made from regenerated cellulose membrane
  • MWCOs from 3500 to 14,000 Daltons
  • Individual lot data sheets from outside laboratory supplied with tubing Types T1, T2, and T3; standard grade Type T4 is not tested in outside laboratory
Description & Specifications
Specifications
0.62 in.
98.42 ft.
T4
Regenerated Cellulose
Two locking nylon membrane clamps, pair of disposable surgical scissors, individual lot data sheets supplied with tubing types T1, T2, and T3
Dialysis Tubing
Diffuses low molecular weight compounds in aqueous solutions, retains proteins and bacteria in tubing
30m
15.9mm
Each
12000 to 14000 d