Fisherbrand™ Regenerated Cellulose Dialysis Tubing
Diffuses low molecular weight compounds in aqueous solutions; retains proteins and bacteria in tubing
 View more versions of this product
Catalog No. 21-152-17
  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Item Description Dialysis Tubing
    Product Type T4
    Quantity Each
    Kit Contents Two locking nylon membrane clamps, pair of disposable surgical scissors, individual lot data sheets supplied with tubing types T1, T2, and T3
    MWCO 12000 to 14000 d
    Diameter (English) 0.25 in.
    For Use With (Application) Diffuses low molecular weight compounds in aqueous solutions, retains proteins and bacteria in tubing
    Length (English) 98.42 ft.
    Length (Metric) 30m
    Diameter (Metric) 6.37mm
    Material Regenerated Cellulose
  • Description & Specifications
Related Products
Product Content Feedback