Learn More
Fisherbrand™ Remote Sensor Module with External Probe for Traceable™ Radio-Signal Hygrometer/Thermometer
For Traceable radio-signal thermometer
Catalog No. 15-077-51
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
This Fisherbrand™ Remote Sensor Module with External Probe is designed for use with the Traceable™ Radio-Signal Remote Thermometer
  • Plastic thermometer probe measuring 0.18 dia. x 0.875 in.L (5 x 22mm)
  • 10 ft. (3m cable) suitable for underwater measurements
Specifications
Specifications
122°F
50°C
-5°C
-5°C to +50°C
0.18 in.
Remote sensor module with external probe for traceable radio-signal thermometer
10 ft.
22 mm
2.2 cm
3 oz.
Radio Signal Remote Thermometer
Remote Sensor Module with External Probe for Traceable Radio-Signal Thermometer
23°F
23°F to 122°F
ISO 17025, A2LA, NIST
0.5 cm
Flip open stand, wall bracket, velcro™ to attach to any surface, two AA and two AAA Alkaline batteries
0.875 in.
3 m
Plastic
0.085 kg