Learn More
Fisherbrand™ Reusable Glass Class A Volumetric Flasks with Standard Taper Stopper Available on GSA/VA Contract for Federal Government customers only.
Single graduation ring blasted on the neck
 View more versions of this product
Catalog No. FB400200
Add to cart
Includes:
  • 41900R ground glass stopper with marking spot
Description
Description
Single graduation ring blasted on the neck
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Compliance
  • Designed from ASTM™ Specification E 288, Class A unserialized requirementsr
Specifications
Specifications
Calibrated To Contain
16
Glass
1.0
Glass
200 mL
Yes
±0.1mL