Learn More
Fisherbrand™ Reusable Glass Rim Top Culture Tubes Available on GSA/VA Contract for Federal Government customers only.
Uniform wall thickness for maximum heat transfer and chemical resistance
 View more versions of this product
Catalog No. FB70016125
Add to cart
Description
Description
  • Made from KG-33 borosilicate glass Durable, white ceramic enamel marking spot
  • Fire-polished top rim
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Non-sterile
16mm
USP Type I, ASTM E 438 Type I, Class A
No Cap
16.25mm
125mm
Borosilicate Glass
19mL
20mL
Yes
19mL
Rim Top
Clear
No
Cell Culture
White Ceramic Enamel Marking Spot