Fisherbrand™ Reusable Glass Tubes with Phenolic Screw Caps Available on GSA/VA Contract for Federal Government customers only.
Phenolic screw cap with white rubber liner
 View more versions of this product
Catalog No. 14-959-25F
  Check Availability
Add to Cart
    Specifications
    Closure Material Phenolic
    For Use With (Application) Suitable for tissue culture work and general bacteriological use
    Label Type Marking Spot
    Liner Material Rubber
    Thread Size 18-415
    Capacity (Metric) 30mL
    Closure Type Screw
    Diameter (Metric) Outer 20mm
    Length (Metric) 150mm
    Material Borosilicate Glass
    Shape Round Bottom
  • Description & Specifications
Related Products
Product Content Feedback