Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Reusable Heavy-Wall Filter Flasks

With heavier walls than standard Erlenmeyer flasks

 View more versions of this product

Catalog No. FB300500



Add to cart

Description

Description

With heavier walls than standard Erlenmeyer flasks

 

  • Side tubulation Capacity scale
  • Designed to take 1 atmosphere of vacuum or 29 in. of mercury
  • Sizes 25 through 500mL have side hose connection designed to accept 5.16 in. O.D. flexible tubing
  • Sizes 1000mL and larger accept 3.8 in. O.D. tubing

 

Specifications

Specifications

ASTM E 1406, Type II, Class I
4.09 in.
10.4cm
Borosilicate Glass
50mL
19cm
7
0.78cm
500 mL
150 to 500mL
7.48 in.
840°F (450°C) max.
Documents
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit