Learn More
Fisherbrand™ RNase Displace™ Decontaminant
Removes RNase and DNA contamination without a time-consuming autoclaving step
This item is not returnable.
View return policy
Description
- Can be used on all surfaces, including glass, plastic, metal, and ceramic
- Works instantly
- Totally rinsable from the decontaminated surface
- Phosphate-free
Specifications
Liquid | |
Milky White | |
<1% Potassium Hydroxide | |
2 to 12 hr. |
13 | |
>212°F | |
All surfaces, including Glass, Plastic, Metal and Ceramic, removes DNase and DNA contamination without a time-consuming autoclaving step | |
1 gal. |