Learn More
Fisherbrand™ Robotic Screw Top Autosampler Vial Closures, Closure with PTFE/Silicone Septa Available on GSA/VA Contract for Federal Government customers only.
For use with Fisherbrand™ Robotic Screw Top Autosampler Vials (cat. no. 033918).
 View more versions of this product
Catalog No. 03-391-31
  Check Availability
Add to cart
Description
Description
For use with Fisherbrand™ Robotic Screw Top Autosampler Vials (cat. no. 033918).
  • Blue closures are preassembled with a 9mm thread and choice of PTFE/Silicone, PTFE/Silicone/PTFE, or PTFE/Red Rubber septa
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Closure
PTFE/Red Rubber
Snap cap
9mm
Blue
100/Pk.