Learn More
Fisherbrand™ Rubber Bulb-Type Safety Pipet Fillers
Tough 100% natural rubber patented pipet fillers eliminate mouth pipetting and handling of hazardous materials
$76.00
Specifications
Autoclavable Not Autoclavable
Type Pipet Filler
For Use With (Equipment) Pipet
Material Natural Rubber
View More Specs
Products 2
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
13-681-50 Edge
Accessories View Documents
Black
Each for $76.00
Case of 72 EA for $5,472.00
Edge
Add to cart
13-681-51 Edge
Accessories View Documents
Red
Each for $76.00
Case of 72 EA for $5,472.00
Edge
Add to cart
Description
Description
Tough 100% natural rubber patented pipet fillers eliminate mouth pipetting and handling of hazardous materials
Two stainless-steel ball valves control vacuum for precise pipet filling and liquid delivery with one hand.
  • Expel air by squeezing bulb, then draw up liquid by pressing valve “↑” on stem
  • To release, press valve “↓” on side of stem
  • Fit pipets with O.D. ranging from 0.187 to 0.375 in. (4.5 to 9mm)
  • Flip style pipette fillers enables one handed bulb operation and removal of top white cap to clean out the bulb from inside
Specifications
Specifications
Not Autoclavable
Pipet Filler
Pipet
Natural Rubber