Fisherbrand™ Safety Blue Thermohydrometer, Celsius Available on GSA/VA Contract for Federal Government customers only.
 View more versions of this product
Catalog No. 11-555-215
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) For determining the percentage by weight of sucrose
    Brix Range 59 to 71
    Ballast Type Lead and mercury-free
    Certifications/Compliance NIST
  • Description & Specifications
Related Products
Product Content Feedback