Fisherbrand™ Safety Coated Amber Boston Round Bottle with White Polypropylene Pulp/Vinyl Cap Available on GSA/VA Contract for Federal Government customers only.

Ideal lab bottle for handling volatile substances. Fisherbrand™ Safety Coated Amber Boston Round Bottle with White Polypropylene Pulp/Vinyl Cap is easier to grip and harder to break.

Catalog No. 02-911-897

  Check Availability
Add to Cart

    Specifications

    Capacity 4 oz., 120mL
    Product Type Safety Coated Boston Round Bottle
    Closure Color White
    Closure Type Screw
    Liner Material Pulp/Vinyl
    Mouth Narrow
    Neck Finish 22-400
    Quantity 24/Cs.
    Shape Boston Round
    Closure Material Polypropylene
    Capacity (Metric) 4 oz., 120mL
    Material Safety Coated Glass
    Capacity (English) 4 oz.
    Closure Screw
    Closure Liner Poly/Vinyl
    Closure Size 22-400
    Color Amber

    • Boston Rounds that have been coated in a layer of Plastisol
    • If the glass does break, the plastisol coating contains substance in the bottle long enough to allow for proper disposal
    • Feature a round body, curved shoulders, and a narrow screw neck opening
    • Amber color offers UV protection and chemical resistance

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit