Fisherbrand™ Safety Coated Amber Boston Round Bottles with White Polypropylene PE Foam/PTFE Lined Caps Available on GSA/VA Contract for Federal Government customers only.

Ideal lab bottles for handling volatile substances. Fisherbrand™ Safety Coated Amber Boston Round Bottles with White Polypropylene PE Foam/PTFE Lined Cap offer chemical resistance and UV protection.

 View more versions of this product

Catalog No. 02-911-894

  Check Availability
Add to Cart

    Specifications

    Capacity (Metric) 120mL
    Capacity 4 oz., 120mL
    Closure Included Yes
    Product Type Safety Coated Boston Round Bottle
    Closure Color White
    Closure Type Screw
    Autoclavable No
    Liner Material PE Foam/PTFE
    Mouth Narrow
    Mouth Style Narrow
    Neck Finish 22-400
    Quantity 24/Cs.
    Shape Boston Round
    Closure Material Polypropylene
    Material Safety Coated Glass
    Capacity (English) 4 oz.
    Closure Screw
    Closure Size 22-400
    Color Amber
    Closure Liner PTFE

    • Plastic-coated Boston Round bottles are easier to grip and harder to break than ordinary glass bottles
    • If the glass does break, the plastisol coating contains the substance in the bottle long enough to allow for proper disposal
    • Features a round body, curved shoulders, and a narrow screw neck opening

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit