Learn More
Fisherbrand™ Safety Coated Amber Wide Mouth Packer Bottles without Cap
General use bottles ideal when UV protection is important. Fisherbrand™ Safety Coated Amber Wide Mouth Packer Bottles without Cap absorbs all wavelengths in the ultraviolet spectrum.
 View more versions of this product
Catalog No. 02-911-906
  Check Availability
Add to cart
Description
Description
Safety Coated Amber Wide Mouth Packer Bottles are normal glass packers coated in plastisol, which is a solution of PVC resin in a liquid plasticizer that cools to form a perfectly-fitting plastic coating. If the glass breaks, the plastic coating contains the substance in the bottle long enough to allow for proper disposal. Plus, these plastic coated glass packer bottles are harder to break and easier to grip than normal glass bottles, reducing the likelihood of breakage occurring in the first place. Amber Glass Packer Bottles feature a wide mouth and are a great general purpose bottle for solids. The amber color filters out ultraviolet wavelengths, the glass offers excellent chemical resistance with minimal chemical interference, and the plastisol coating offers additional protection against lab mishaps.
  • Features narrow screw neck opening, curved shoulders, and a round body
  • Offers minimal chemical interference with contents
  • Ensure integrity for long term storage and protection for UV-sensitive contents
Specifications
Specifications
16 oz., 500mL
16 oz.
Safety Coated GlassþPlastic Coated Glass
Wide Mouth Packer Bottle
53-400
Screw
500mL
AmberþAmber
12/PK.
No
Wide
Documents
Promotions
Promotions
  • Get the essentials you need at a price you'll love.
    12-31-2018
  • Fisherbrand , Fisher Scientific , DWK Life Sciences (Kimble)
    Get up to $250 in glassware at no additional cost with your qualifying Fisherbrand™ Glassware purchase.
    06-30-2019