Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Safety Coated Amber Wide Mouth Packer Bottles with Polypropylene PE Foam/PTFE Lined Cap

Ideal general purpose bottle for solids. Fisherbrand™ Safety Coated Amber Wide Mouth Packer Bottles with Polypropylene PE Foam/PTFE Lined Cap filters out ultraviolet wavelengths for light sensitive solids.

 View more versions of this product

Catalog No. 02-911-831


  Check Availability
Add to cart

Specifications

Description

  • Normal glass packers coated in plastisol, which is a solution of PVC resin in liquid plasticizer that cools to form perfectly-fitting plastic coating
  • Plastic coating contains substance in bottle long enough to allow for proper disposal if glass breaks
  • Easier to grip than normal glass bottles
Description & Specifications

Specifications

32 oz., 1000mL
32oz.
Polyethylene
Safety Coated Glass
Bottle, Safety Coated Wide Mouth Packer
White
PE Foam/PTFE
53-400
Screw
960mL
Polypropylene
Amber
12/Cs.
Yes
53-400
Wide
Round
Promotions

Promotions

  • Fisherbrand
    For a limited time, save 35% on the price of Fisherbrand™ Safety Coated Amber Wide Mouth Packer Bottles.

    07-01-2017

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit