Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Safety Coated Amber Wide Mouth Packer Bottles with White Polypropylene PE Foam/PTFE Cap

Provides excellent storage for liquids and solids in the laboratory. Fisherbrand™ Wide Neck Soda Lime Glass Bottles are to be sealed with waterproof screw caps.

 View more versions of this product

Catalog No. 02-911-863


  Check Availability
Add to cart

Description

Description

  • Normal glass packers coated in plastisol, which is a solution of PVC resin in liquid plasticizer that cools to form perfectly-fitting plastic coating
  • Plastic coating contains substance in bottle long enough to allow for proper disposal if glass breaks
  • Easier to grip than normal glass bottles
Specifications

Specifications

8 oz., 250mL
8.5oz.
Polyethylene
Safety Coated GlassþPlastic Coated Glass
Bottle, Safety Coated Wide Mouth PackerþWide Mouth Packer Bottle
White
PE Foam/PTFE
45-400
screw cap
250mL
Polypropylene
AmberþAmber
24/Cs.
Yes
45-400
Wide
Round
Documents
Promotions

Promotions

  • Fisherbrand , Fisher Scientific , DWK Life Sciences (Kimble)
    Get up to $250 in glassware at no additional cost with your qualifying Fisherbrand™ Glassware purchase.

    06-30-2019

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit