Fisherbrand™ Safety Coated Amber Wide Mouth Packer Bottles with Polypropylene PE Foam/PTFE Lined Cap Available on GSA/VA Contract for Federal Government customers only.

Ideal general purpose bottle for solids. Fisherbrand™ Safety Coated Amber Wide Mouth Packer Bottles with Polypropylene PE Foam/PTFE Lined Cap filters out ultraviolet wavelengths for light sensitive solids.

 View more versions of this product

Catalog No. 02-911-830

  Check Availability
Add to Cart

    Specifications

    Closure Included Yes
    Product Type Safety Coated Wide Mouth Packer Bottle
    Closure Color White
    Closure Type Screw
    Liner Material PE Foam/PTFE
    Mouth Wide
    Mouth Style Wide
    Neck Finish 53-400
    Quantity 12/Cs.
    Shape Round
    Capacity (Metric) 16 oz., 500mL
    Closure Attached cap
    Closure Material Polypropylene
    Material Safety Coated Glass
    Capacity (English) 16 oz.
    Closure Liner Polyethylene
    Closure Size 53-400
    Color Amber
    Capacity 16 oz., 500mL

    • Normal glass packers coated in plastisol, which is a solution of PVC resin in liquid plasticizer that cools to form perfectly-fitting plastic coating
    • Plastic coating contains substance in bottle long enough to allow for proper disposal if glass breaks
    • Easier to grip than normal glass bottles

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit