Fisherbrand™ Safety Coated Clear Boston Round Bottles with White Polypropylene Pulp/Vinyl Cap

Ideal lab bottle for handling volatile substances. Fisherbrand™ Safety Coated Clear Boston Round Bottles with White Polypropylene Pulp/Vinyl Cap are easier to grip and harder to break.

 View more versions of this product

Catalog No. 02-911-818

  Check Availability
Add to Cart

    Specifications

    Product Type Safety Coated Boston Round Bottle
    Closure Color White
    Closure Type Screw
    Liner Material Pulp/Vinyl
    Mouth Narrow
    Neck Finish 24-400
    Quantity 12/Cs.
    Shape Boston Round
    Capacity (Metric) 8oz., 240mL
    Capacity (English) 8oz.
    Closure Material Polypropylene
    Material Safety Coated Glass
    Closure Screw
    Closure Liner Poly/Vinyl
    Closure Size 24-400
    Color Clear
    Capacity 8 oz., 250mL

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit