Learn More
Fisherbrand™ Safety Coated Clear Boston Round Bottles with Black Phenolic Rubber Cap
Ideal lab bottle for handling volatile substances. Fisherbrand™ Clear Boston Round Bottles with Black Phenolic Rubber Cap are easier to grip and harder to break.
 View more versions of this product
Catalog No. 02-911-910
Add to cart
Includes: Quantity,ClosureIncluded,ClosureLiner,ClosureMaterial,ClosureType,dimColor,dimMaterial,Autoclavable,dimSterility,dimType
Description
Description
Safety Coated Glass Boston Round Bottles are glass Boston Rounds that have been coated in a layer of Plastisol, a solution of PVC resin in a liquid plasticizer that cools to form a perfectly-fitting plastic coating. These plastic-coated glass Boston Round bottles are easier to grip and harder to break than ordinary glass bottles, yet still offer the chemical resistance and minimal interference of glass. In the case that the glass does break, the plastisol coating will contains the contents in the bottle long enough to allow for proper disposal. Boston Round bottles feature a round body, curved shoulders, and a narrow screw neck opening. Safety plastic-coated Boston Round bottles are ideal lab bottles for handling volatile substances.
  • Boston Rounds that have been coated in a layer of Plastisol
  • If the glass does break, the plastisol coating contains substance in the bottle long enough to allow for proper disposal
  • Features a round body, curved shoulders, and a narrow screw neck opening
  • Not autoclavable
Order Info
Quantity,ClosureIncluded,ClosureLiner,ClosureMaterial,ClosureType,dimColor,dimMaterial,Autoclavable,dimSterility,dimType
Specifications
Specifications
8 oz., 250mL
8 oz.
Rubber
Glass, Plastic Coated Glass
24-400
250mL
Rubber
Clear, Clear
Boston Round Bottle
Screw