Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Safety-Labeled Right-to-Know Wash Bottles Available on GSA/VA Contract for Federal Government customers only.

Keep liquids in clearly marked bottles that help prevent accidental misuse. Fisherbrand™ Safety-Labeled Right-to-Know Wash Bottles are available in both preprinted and “Label Your Own” bottles.

 View more versions of this product

Catalog No. 08-647-718


Add to cart

Description

Description

 

  • Feature a wide mouth 53mm (2 in.) for easy refilling
  • Bottles are pre-printed with the Chemical name and formula, the National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment
  • Translucent LDPE bottles; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm
  • Color-coded polypropylene closures aid in quick and easy identification
  • Note: Use LYOB only with solvents compatible with LDPE

 

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

32 oz., 1000mL
32 oz.
Yellow, White
LDPE
No
Wide
1000mL
Polypropylene
Sodium Hypochlorite
4/Pk.
Yellow
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit