Fisherbrand™ Safety Lancets
Built-in needle control depth (1.8mm) for consistent and accurate sampling results
 View all options for this product
Catalog No. 23-306-760
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Sterile Yes
    Color Yellow
    Quantity 100/Pk.; 48/Pk./Cs.
    • Sterile lancet needle is hidden inside the cartridge and automatically retracts after use
    • Helps protect patient and health professional against accidental needlestick injuries and cross-contamination
    • Proprietary spring technology inhibits access to lancet after use — lancet cannot be reloaded or reused
  • Description & Specifications
Related Products
Product Content Feedback