Learn More
Fisherbrand™ Safety Lancets
Built-in needle control depth (1.8mm) for consistent and accurate sampling results
 View more versions of this product
Catalog No. 23-306-761
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Green
1.8mm
Helps protect patient and health professional against accidental needlestick injuries and cross-contamination
100 Pack
Retract
0.07 in.
Normal
Proprietary spring technology
Yes
Safety Lancet