Fisherbrand™ Safety Lancets

Built-in needle control depth (1.8mm) for consistent and accurate sampling results

 View more versions of this product

Catalog No. 23-306-762

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Color Blue
    Depth (English) Needle 0.07 in.
    Depth (Metric) Needle 1.8mm
    Flow Type Low
    For Use With (Application) Helps protect patient and health professional against accidental needlestick injuries and cross-contamination
    Includes Proprietary spring technology
    Quantity 100 Pack
    Sterile Yes
    Style Retract
    Product Type Safety Lancet

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit