Fisherbrand™ Sav-It™ Tube Closures Available on GSA/VA Contract for Federal Government customers only.

Firm, leak-resistant seals protect personnel and samples

 View more versions of this product

Catalog No. 02-707-1

  Check Availability
Add to Cart

    Specifications

    Sterile No
    Size (Metric) 16mm
    Material Elastomer Co-polymer
    Color Red
    For Use With (Equipment) Test tubes, centrifuge tubes, specialty tubes, evacuated blood drawing tubes
    Size (English) 0.62 in.
    Autoclavable No

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit