Fisherbrand™ Scientific Technical Pens

Fast-drying ink in four colors

 View more versions of this product

Catalog No. 13-383-1B

  Check Availability
Add to Cart

    Specifications

    Product Type Technical Pen
    Disposable Yes
    For Use With (Application) Used for record results in lab notebooks, fill in charts, draw graphs, mark recording paper, and produce diagrams
    Width (Metric) Line 0.4mm
    Color Black
    Quantity 6 Pack

    Fisherbrand™ Scientific Technical Pens produce extra-fine lines with accuracy—and without smearing. Special “long-legged” point is perfect for writing on notebooks, forms, graphs, charts, tables and recording paper. Fast-drying ink in four colors.

    • Ink is smooth flowing, fadeproof, and quick drying
    • Assortment includes red, blue, and special ultrablack for excellent reproduction on office copiers
    • Large ink reservoir lets you write for over 0.50 mile (800m)
    • Cap design and color indicate point size and color of pen
    • Built-in super-tight cap seal keeps ink from drying out when pen is not in use
    • Choose from two line sizes: extra fine, 0.016 in. (0.4mm), or super fine, 0.008 in. (0.2mm)
    • Make precise lines with T-squares, rulers and flexible curves

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit