Fisherbrand™ Scinti-Rack™ System: Plates

Ideal for transport and storage of vials

 View more versions of this product

Catalog No. 03-337-44

  Check Availability
Add to Cart

    Specifications

    Positions 50.0,50.0
    For Use With (Equipment) Fisherbrand Scinti-Rack™ system
    Autoclavable No,No
    Product Type Rack vial
    Material HDPE
    Type Rack vial
    Color White

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit