Fisherbrand™ Screw Caps for Disposable Glass Tubes Available on GSA/VA Contract for Federal Government customers only.

Phenolic caps have cemented-in rubber liners

 View more versions of this product

Catalog No. 14-957-82A

  Check Availability
Add to Cart

    Specifications

    Color Black
    Liner Material Rubber
    Product Type Screw Cap
    For Use With (Equipment) Disposable glass centrifuge tubes
    GPI Thread 13-415
    Thread Size 13-415
    Autoclavable Yes
    Material Phenolic

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit