Fisherbrand™ Sharps-A-Gator™ Point-of-Use Sharps Containers

Horizontal loading, nonmechanical sharps opening provides one-hand, one-step, no-touch sharps depositing

 View all options for this product

Catalog No. 14-827-62


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Autoclavable Yes
    Capacity (English) 2.5 qt.
    Capacity (Metric) 2.4L
    Closure Type Dual Opening Hinge w/ VAT Perimeter Locks; Temporary
    Height (English) 7.5 in.
    Height (Metric) 19cm
    Material Puncture-resistant Polypropylene (PP)
    Length (English) 4.2 in.
    Length (Metric) 11cm
    Quantity 30/cs.
    Width (English) 11 in.
    Width (Metric) 28cm
    Color Red
    Capacity 2.5 qt., 2.4L
    Dimensions (L x W x H) 4.2 x 11 x 7.5 in. (11 x 28 x 19cm)

    • For use in patient rooms and all patient-care areas Opening design impedes hand entry to reduce risk of needle sticks when disposing of sharps
    • Clear tops and translucent containers allow you to monitor fill level
    • Puncture-resistant, durable polypropylene Can be autoclaved or incinerated
    • Clearly marked with biohazard label

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit