Fisherbrand™ SHUR/Clean™ Passive Open-Front Fume Hoods Accessories

 View more versions of this product

Catalog No. 22-047-212

  Check Availability
Add to Cart

    Specifications

    Depth (Metric) Exterior 56cm
    For Use With (Application) 48 in. fume hood
    Height (Metric) Exterior 12cm
    Width (Metric) Exterior 69cm
    Depth (English) Exterior 22 in.
    Height (English) Exterior 12 in.
    Type Reagent Bowl Shelf
    Width (English) Exterior 27 in.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit