Fisherbrand™ Silicone Bath Fluids
Provide thermal stability over a variety of temperature ranges
 View more versions of this product
Catalog No. 13-874-60D
  Check Availability
Add to Cart
    Specifications
    Product Type Silicone bath fluid
    Freezing Point -22°C
    Fluid Delivery Type DC710
    Material Silicone
    Temperature (Metric) Range 150° to 250°C
    Viscosity 500 Cs.
    Capacity (English) 1 gal.
  • Description & Specifications
Related Products
Product Content Feedback