Learn More
Fisherbrand™ Silicone Bulb-Type Safety Pipet Fillers
Chemical-resistant, resilient, silicone pipet fillers with glass valves improve worker safety
$41.55
Specifications
Autoclavable Not Autoclavable
Type Pipet Filler
For Use With (Equipment) Pipet
Material Silicone
View More Specs
Products 2
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
13-681-102B Edge
Accessories View Documents
Black
Each for $41.55
Edge
Add to cart
13-681-102A Edge
Accessories View Documents
Red
Each for $41.55
Edge
Add to cart
Description
Description
Improve worker safety with these chemical-resistant, resilient, silicone pipet fillers with glass valves.
  • Eliminate mouth pipetting and handling of hazardous materials
  • Fill and dispense pipets with one hand
  • Long-lasting silicone is highly resistant to organic solvents
  • Fit pipets with O.D. ranging from 0.187 to 0.375 in. (4.5 to 9mm)
Specifications
Specifications
Not Autoclavable
Pipet Filler
Pipet
Silicone