Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Silicone Bulb-Type Safety Pipet Fillers
Chemical-resistant, resilient, silicone pipet fillers with glass valves improve worker safety
Description
Improve worker safety with these chemical-resistant, resilient, silicone pipet fillers with glass valves.
- Glass valves improve worker safety by eliminating mouth pipetting and handling of hazardous materials
- Long-lasting silicone is highly resistant to organic solvents
- Fit any pipet with O.D. ranging from 0.19 to 0.38 in. (4.5 to 9 mm)
To Use:
- Fill and dispense pipets with one hand
- Expel air by squeezing valve '1' above bulb
- Draw up liquid by pressing valve '2' on stem
- To release, press valve '3' on side of stem
Specifications
No | |
Not Autoclavable | |
Pipette | |
3 Valve Standard | |
Pipette Filler | |
Pipet |
4.5 mm to 9 mm | |
Black | |
Silicone | |
1 | |
50 mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction