Fisherbrand™ Silicone Plate Seals Available on GSA/VA Contract for Federal Government customers only.

Excellent chemical compatibility and resistance to tearing and coring

 View more versions of this product

Catalog No. 03-396-49

  Check Availability
Add to Cart

    Specifications

    Width (Metric) 7mm
    Material Silicone
    No. per Pack 10.0
    Product Type 96 Well Silicone Plate Seal
    Autoclavable Yes
    Pre-slit No
    Applications Excellent chemical compatibility and resistance to tearing and coring
    Color Clear

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit