Learn More
Bio Plas Siliconized Screw-Cap Microcentrifuge Tubes
Ideal for packaging, transporting, and storing diagnostic tests, especially in the area of biotechnology, molecular biological products and reagents
$413.09 - $1465.51
Specifications
Color Natural
Capacity (Metric) 2mL
Material Polypropylene
Max. Force 25000xG
Shape Conical
View More Specs
Products 2
Catalog Number Mfr. No. Closure Type Sterility Price Quantity & Availability  
Catalog Number Mfr. No. Closure Type Sterility Price Quantity & Availability  
05-541-66
View Documents
Bio Plas
4204SL
Without Non-sterile Pack of 1000 for $413.09
Add to cart
05-541-67
View Documents
Bio Plas
4204SLS
Screw Sterile Pack of 1000 for $1,465.51
Add to cart
Description
Description
Low surface adhesion reduces samples adhering to walls.
  • Polypropylene
  • Natural color
  • Conical with skirt
  • Available sterile or nonsterile, with or without screw cap
  • Withstand centrifugal forces up to 25,000xG
  • Sterile model includes tight-seal closure that reduces accidental opening during use
Order Info
1000 per pack, 5 packs per case
Specifications
Specifications
Natural
Polypropylene
Conical
2mL
25000xG