Learn More
Fisherbrand™ Snap-Cap™ Flat-Top Graduated Microcentrifuge Tubes
Available as sterile and nonsterile
 View more versions of this product
Catalog No. 02-682-552
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Available as sterile and nonsterile
  • Polypropylene
  • Graduated
  • In different colors
Order Info
Sterile tubes are individually wrapped to maintain sterilty
Specifications
Specifications
Snap Cap with Flat Top
Flat Snap
Green
39mm
17000xG
Bulk
Non-sterile
1.5mL
14mm
Polypropylene
0.5mL