Learn More
Fisherbrand™ Snap-Cap™ Flat-Top Graduated Microcentrifuge Tubes
Available as sterile and nonsterile
 View more versions of this product
Catalog No. 02-681-255
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
Snap Cap with Flat Top
0.6mL
10mm
Polypropylene
No
Bulk
Snap
Amber
30mm
30000xG
0.2mL