Learn More
Fisherbrand™ Snap-Cap™ Microcentrifuge Tubes: Flat Top
Polypropylene tubes
$136.60
Specifications
Cap Style Snap Cap with Flat Top
Certifications/Compliance RNase, DNase, RNA, DNA, ATP, pyrogen-free
Material Polypropylene
Packaging Bulk
Sterility Non-sterile
View More Specs
Products 8
Catalog Number Capacity (Metric) Closure Type Color Max. Force Price Quantity & Availability  
Catalog Number Capacity (Metric) Closure Type Color Max. Force Price Quantity & Availability  
02-681-240
View Documents
0.6mL Snap Natural 30000xG
Pack of 1000 for $136.60
Case of 10 PK for $1,161.00
Add to cart
02-681-241
View Documents
0.6mL Snap Red 30000xG
Pack of 1000 for $136.60
Case of 10 PK for $1,161.00
Add to cart
02-681-243
View Documents
0.6mL Snap Green 30000xG
Pack of 1000 for $136.60
Case of 10 PK for $1,161.00
Add to cart
02-681-242
View Documents
0.6mL Snap Blue 30000xG
Pack of 1000 for $136.60
Case of 10 PK for $1,161.00
Add to cart
02-681-244
View Documents
0.6mL Snap Yellow 30000xG
Pack of 1000 for $136.60
Case of 10 PK for $1,161.00
Add to cart
02-681-245
View Documents
0.6mL Snap Orange 30000xG
Pack of 1000 for $136.60
Case of 10 PK for $1,161.00
Add to cart
02-681-246
View Documents
0.6mL Snap Violet 30000xG
Pack of 1000 for $136.60
Case of 10 PK for $1,161.00
Add to cart
02-681-248
View Documents
0.6mL Snap Assorted 30000xG
Pack of 1000 for $136.60
Case of 10 PK for $1,161.00
Add to cart
Description
Description
  • Packed in bulk Nonsterile
Specifications
Specifications
Snap Cap with Flat Top
Polypropylene
Non-sterile
RNase, DNase, RNA, DNA, ATP, pyrogen-free
Bulk