Learn More
Fisherbrand™ Solid Silicone Stoppers
Create a tight, reliable seal in virtually all sealing applications.
 View more versions of this product
Catalog No. 09-704-1Y
Add to cart
Description
Description
Create a tight, reliable seal in virtually all sealing applications.
  • Pure, high-grade, translucent silicone withstands repeated sterilization, as well as temperatures from -60° to +220°C (-76° to +428°F) peaks at 250°C (482°F)
  • Resist particle buildup Chemically inert and are food-grade quality
Ideal for procedures demanding high purity—ideal for biological applications (after normal cleaning and autoclaving)
Specifications
Specifications
100D
-60° to 220°C
Silicone
99.8mm
6.5cm
15/100D
Translucent
34mm
Ideal for procedures demanding high purity ideal for biological applications
Solid Stopper