Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Saint-Gobain Solid Silicone Stoppers

Create a tight, reliable seal in virtually all sealing applications.

$5.29 - $226.50

Specifications

Color Translucent
Material Silicone
Product Type Solid Stopper
For Use With (Application) Ideal for procedures demanding high purity ideal for biological applications
Temperature Range (Metric) -60° to 220°C
View More Specs

Products 24
Catalog Number Mfr. No. Length (Metric) Size Stopper Number Price Quantity & Availability  
Catalog Number Mfr. No. Length (Metric) Size Stopper Number Price Quantity & Availability  
09-704-1
View Documents
Saint-Gobain
D1069804
1.5cm 6D 6D
Pack of 10 for $10.12
Add to cart
 
09-704-1B
View Documents
Saint-Gobain
D1069806
2cm 10D 10D
Pack of 10 for $14.49
Add to cart
 
09-704-1A
View Documents
Saint-Gobain
D1069805
2cm 8D 8D
Pack of 10 for $13.04
Add to cart
 
09-704-1C
View Documents
Saint-Gobain
D1069807
2cm 13D 000/13D
Pack of 10 for $16.61
Add to cart
 
09-704-1D
View Documents
Saint-Gobain
D1069808
2cm 15D 00/15D
Pack of 10 for $5.29
Add to cart
 
09-704-1G
View Documents
Saint-Gobain
D1069811
2.5cm 21D 2/21D
Pack of 10 for $38.95
Add to cart
 
09-704-1K
View Documents
Saint-Gobain
DX26303110
3cm 31D 6/31D
Pack of 10 for $119.70
Add to cart
 
09-704-1F
View Documents
Saint-Gobain
D1069810
2cm 18D 1/18D
Pack of 10 for $33.45
Add to cart
 
09-704-1E
View Documents
Saint-Gobain
D1069809
2cm 16D 0/16D
Pack of 10 for $26.85
Add to cart
 
09-704-1L
View Documents
Saint-Gobain
D1069816
3cm 34D 6.5/34D
Pack of 10 for $115.30
Add to cart
 
09-704-1H
View Documents
Saint-Gobain
D1069812
3cm 24D 3/24D
Pack of 10 for $50.30
Add to cart
 
09-704-1J
View Documents
Saint-Gobain
DX26302910
3cm 29D 5/29D
Pack of 10 for $84.70
Add to cart
 
09-704-1M
View Documents
Saint-Gobain
D1069817
3.5cm 38D 7/38D
Pack of 10 for $140.80
Add to cart
 
09-704-1N
View Documents
Saint-Gobain
D1069818
3.5cm 41D 8/41D
Pack of 10 for $162.10
Add to cart
 
09-704-1Q
View Documents
Saint-Gobain
D1069820
4cm 49D 10/49D
Pack of 5 for $146.90
Add to cart
 
09-704-1P
View Documents
Saint-Gobain
D1069819
4cm 45D 9/45D
Pack of 10 for $226.50
Add to cart
 
09-704-1T
View Documents
Saint-Gobain
D1069823
4.5cm 65D 11.5/165D
Each for $51.30
Add to cart
 
09-704-1S
View Documents
Saint-Gobain
D1069822
4.5cm 60D 11/60D
Pack of 5 for $209.50
Add to cart
 
09-704-3
View Documents
Saint-Gobain
D1069824
5cm 70D 13/70D
Each for $62.60
Add to cart
 
09-704-1I
View Documents
Saint-Gobain
DX26302710
3cm 27D 4/27D
Pack of 10 for $60.90
Add to cart
 
09-704-1W
View Documents
Saint-Gobain
D1069826
6cm 83 83D This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
09-704-1Y
View Documents
Saint-Gobain
DX26310001
6.5cm 100D 15/100D This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
09-704-1R
View Documents
Saint-Gobain
D1069821
4cm 54D 10.5/54D This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
09-704-1V
View Documents
Saint-Gobain
D1069825
5.5cm 75D 75D This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Create a tight, reliable seal in virtually all sealing applications.

 

  • Pure, high-grade, translucent silicone withstands repeated sterilization, as well as temperatures from -60° to +220°C (-76° to +428°F) peaks at 250°C (482°F)
  • Resist particle buildup
  • Chemically inert and are food-grade quality

 

Ideal for procedures demanding high purity—ideal for biological applications (after normal cleaning and autoclaving).

Specifications

Specifications

Translucent
Solid Stopper
-60° to 220°C
Silicone
Ideal for procedures demanding high purity ideal for biological applications
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit