Learn More
Fisherbrand™ Solid Silicone Stoppers
Create a tight, reliable seal in virtually all sealing applications.
 View more versions of this product
Catalog No. 09-704-1D
Add to cart
Description
Description
Create a tight, reliable seal in virtually all sealing applications.
  • Pure, high-grade, translucent silicone withstands repeated sterilization, as well as temperatures from -60° to +220°C (-76° to +428°F) peaks at 250°C (482°F)
  • Resist particle buildup Chemically inert and are food-grade quality
Ideal for procedures demanding high purity—ideal for biological applications (after normal cleaning and autoclaving)
Specifications
Specifications
10.5mm
Ideal for procedures demanding high purity ideal for biological applications
Solid Stopper
00/15D
Translucent
14mm
2cm
15D
-60° to 220°C
Silicone