Fisherbrand™ Solvent-Vented™ and Safety-Labeled Wash Bottles Available on GSA/VA Contract for Federal Government customers only.

Are pre-printed with Chemical name and formula. Bel Art™ Solvent-Vented™ and Safety-Labeled Wash Bottles have patented valve eliminates drips and allows bottle to dispense upright or inverted.

 View more versions of this product

Catalog No. 03-409-37

  Check Availability
Add to Cart

    Specifications

    Capacity (English) 16 oz.
    Certifications/Compliance OSHA 29 CFR 1910.1200
    Closure Material Polypropylene
    Color Translucent
    Material LDPE
    Mouth Wide
    Capacity (Metric) 500mL
    Autoclavable No
    For Use With Acetone
    Capacity 16 oz., 500mL

    • Wide mouth 2 in. (53mm) bottle for easy refilling
    • Polypropylene closure with leakproof design, color-coded to aid in easy identification
    • Translucent, LDPE solvent-resistant; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm; Toluene is red LDPE bottle
    • Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
    • Labeled with six of the most commonly used solvent names or label your own; custom labeling available

     

    Use LYOB only with solvents compatible with LDPE.


  • Description & Specifications
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit