Fisherbrand SQ-EASY™ PreFilter Tips
Disposable, efficient self-contained filtration system for sample preparation applications
 View more versions of this product
Catalog No. 22-267-392
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) For use in stool, Serum, diluted Whole Blood, Urine, and throat swab assays
    For Use With (Equipment) PreFilter™ system
    Particle Retention 25μm
    Quantity 100 Pack
    Tip Type Hydrophilic filter
    Material HDPE
    Product Type Sq-Easy™ Filter Tip
  • Description & Specifications
Related Products
Product Content Feedback